missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde, 40 wt.% solution in water, ACROS Organics™

Brand:  Acros Organics 175790025

Code : 29

Additional Details : CAS Number : 78-98-8 Weight : 4.00000kg


Quantity 2.5kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10606594

  • £495.00 / 2.50kg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Pyruvic aldehyde
7732-18-5
C3H4O2
72.06
Pyruvaldehyde
35-45 wt.%
35-45 wt.%
CH3COCHO
MFCD00006960

Specifications

Pyruvic aldehyde
total acid (HCOOH): < 7.0%
7732-18-5
1.4209
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, benzene and dmso
1.2
CH3COCHO
MFCD00006960
1.2000g/mL
Pyruvaldehyde
1 to 3
> 72.0°C
2.5kg
35-45 wt.%
Glass bottle
Solution
35-45 wt.%
C3H4O2
72.06
01, 762
15, 6153
Clear yellow to yellow-brown
− 20.0°C
>71°C
Videos
SDS