missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde, 35-45% w/w aq. soln, Thermo Scientific Chemicals

Catalog Number p-7027698
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
78-98-8
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
This item is not returnable. View return policy
78-98-8
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O

Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

Solubility
Soluble in DMSO, water.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Density 1.2
Linear Formula CH3COCHO
Refractive Index 1.405
Quantity 25 g
Beilstein 906750
Sensitivity Air Sensitive
Merck Index 14,6081
Solubility Information Soluble in DMSO,water.
Formula Weight 72.06
Concentration or Composition (by Analyte or Components) 35 to 45% w/w aq. soln
Chemical Name or Material Pyruvic aldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'