missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 11926611
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
109-97-7
C4H5N
67.09
MFCD00005216
KAESVJOAVNADME-UHFFFAOYSA-N
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
8027
CHEBI:19203
1H-pyrrole
N1C=CC=C1
109-97-7
C4H5N
67.09
MFCD00005216
KAESVJOAVNADME-UHFFFAOYSA-N
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
8027
CHEBI:19203
1H-pyrrole
N1C=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -23.0°C
Color Brown to Colorless
Density 0.9600g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100
Quantity 2.5 kg
Beilstein 20,159
Merck Index 15,8128
Specific Gravity 0.96
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents
Viscosity 1.317 mPa.s (20°C)
Formula Weight 67.09
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyrrole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'