missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrole-3-carboxylic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
Solubility
Soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
Solubility
Soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
Specifications
Quantity | 250 mg |
Beilstein | 108642 |
Solubility Information | Soluble in water. |
Formula Weight | 111.1 |
Percent Purity | ≥98% |
Chemical Name or Material | Pyrrole-3-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'