missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxaldoxime, 97%, Thermo Scientific™

Catalog Number p-7047359
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
32597-34-5
C5H6N2O
110.116
MFCD00965095
WJMIDGRLWRUSSN-UHFFFAOYSA-N
1h-pyrrole-2-carbaldehyde oxime, 1h-pyrrole-2-carboxaldehyde oxime, n-1h-pyrrol-2-ylmethylidene hydroxylamine, e-n-1h-pyrrol-2-ylmethylidene hydroxylamine, n-pyrrol-2-ylidenemethyl hydroxylamine, z-n-1h-pyrrol-2-ylmethylidene hydroxylamine
6743783
N-(pyrrol-2-ylidenemethyl)hydroxylamine
C1=CC(=CNO)N=C1
This item is not returnable. View return policy
32597-34-5
C5H6N2O
110.116
MFCD00965095
WJMIDGRLWRUSSN-UHFFFAOYSA-N
1h-pyrrole-2-carbaldehyde oxime, 1h-pyrrole-2-carboxaldehyde oxime, n-1h-pyrrol-2-ylmethylidene hydroxylamine, e-n-1h-pyrrol-2-ylmethylidene hydroxylamine, n-pyrrol-2-ylidenemethyl hydroxylamine, z-n-1h-pyrrol-2-ylmethylidene hydroxylamine
6743783
N-(pyrrol-2-ylidenemethyl)hydroxylamine
C1=CC(=CNO)N=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 163°C to 166°C
Quantity 1 g
Beilstein 108674
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material Pyrrole-2-carboxaldoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'