Learn More
Pyroxamide, Tocris Bioscience™
Histone deacetylase inhibitor
Brand:  RD Systems (Bio-Techne) 4403/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13434704
  • £169.05 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Inhibitor of histone deacetylase (HDAC); potently inhibits affinity purified HDAC1. Also inhibits the growth of tumor cells in vitro and in vivo. Induces p21/WAF1 expression in tumor cells.
Specifications
Chemical Identifiers
382180-17-8
265.313
pyroxamide, n-hydroxy-n'-3-pyridinyloctanediamide, n1-hydroxy-n8-pyridin-3-yl octanediamide, n-hydroxy-n'-pyridin-3-yl octanediamide, unii-12n86dss23, polar hybrid compound, octanediamide, n-hydroxy-n'-3-pyridinyl, suberoyl-3-aminopyridineamide hydroxamic acid, n-hydroxy-n'-3-pyridyl-1,8-octanediamide, 8-hydroxyamino-8-oxo-n-3-pyridyl octanamide
N'-hydroxy-N-pyridin-3-yloctanediamide
C13H19N3O3
PTJGLFIIZFVFJV-UHFFFAOYSA-N
4996
C1=CC(=CN=C1)NC(=O)CCCCCCC(=O)NO
Specifications
10mg
382180-17-8
pyroxamide, n-hydroxy-n'-3-pyridinyloctanediamide, n1-hydroxy-n8-pyridin-3-yl octanediamide, n-hydroxy-n'-pyridin-3-yl octanediamide, unii-12n86dss23, polar hybrid compound, octanediamide, n-hydroxy-n'-3-pyridinyl, suberoyl-3-aminopyridineamide hydroxamic acid, n-hydroxy-n'-3-pyridyl-1,8-octanediamide, 8-hydroxyamino-8-oxo-n-3-pyridyl octanamide
C1=CC(=CN=C1)NC(=O)CCCCCCC(=O)NO
265.313
265.31
Pyroxamide
C13H19N3O3
PTJGLFIIZFVFJV-UHFFFAOYSA-N
N'-hydroxy-N-pyridin-3-yloctanediamide
4996
>98%