missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyronin Y, pure, high purity biological stain

Catalog Number 10624902
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
92-32-0
C17H19ClN2O
302.802
MFCD00011725
INCIMLINXXICKS-UHFFFAOYSA-M
C.I. 45005, Pyronin G
7085
CHEBI:87347
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
This item is not returnable. View return policy
92-32-0
C17H19ClN2O
302.802
MFCD00011725
INCIMLINXXICKS-UHFFFAOYSA-M
C.I. 45005, Pyronin G
7085
CHEBI:87347
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorption Ratio 1.23 to 1.7
Dye Content 50% min.
Beilstein 18, V,10, 181
Merck Index 15, 8119
Infrared Spectrum Authentic
Lambda Max 546 to 551nm (in 50% ethanol)
Formula Weight 302.79
Grade Pure
Packaging Glass bottle
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Pyronin Y, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Pyronin Y
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'