Learn More
Lonza Pyrogen-free Test Tubes
Recommended for use as reaction tubes in gel clot assays. Lonza™ Pyrogen-free Test Tubes Product are made from USP Type 1 flint borosilicate glass and contain less than 0.005 EU/ml endotoxin.
Brand: Lonza N201
Code : NEW
Additional Details : Weight : 0.27000kg
Description
- Made from USP Type 1 flint borosilicate glass
- Contain less than 0.005 EU/ml endotoxin
- Recommended for use as reaction tubes in gel clot assays.
Specifications
Test Tube | |
Screw Cap | |
75 mm | |
Borosilicate Glass |