missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-189816
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
87-66-1
C6H6O3
126.11
MFCD00002192
WQGWDDDVZFFDIG-UHFFFAOYSA-N
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
1057
CHEBI:16164
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O
87-66-1
C6H6O3
126.11
MFCD00002192
WQGWDDDVZFFDIG-UHFFFAOYSA-N
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
1057
CHEBI:16164
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 131.0°C to 135.0°C
Color White
Boiling Point 309.0°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula C6H3(OH)3
Quantity 100 g
UN Number 2811
Beilstein 06, 1071
Merck Index 15, 8112
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide
Formula Weight 126.11
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Pyrogallol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyrogallol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'