missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridinium trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
96.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
It is miscible with water, soluble in polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
It is miscible with water, soluble in polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 221°C to 223°C |
Quantity | 25 g |
Solubility Information | It is miscible with water,soluble in polar solvents. |
Formula Weight | 229.17 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Pyridinium trifluoromethanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'