missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine N-oxide, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Pyridine N-oxide is used in the treatment of Kayser's disease. It is widely used in chemical industry. It can act as catalytic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Soluble in water.
Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Specifications
Melting Point | 61°C to 66°C |
Boiling Point | 270°C |
Flash Point | 143°C (289°F) |
Quantity | 25 g |
Beilstein | 105257 |
Sensitivity | Very Hygroscopic |
Merck Index | 14,7972 |
Solubility Information | Soluble in water. |
Formula Weight | 95.1 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'