missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals

Catalog Number p-189656
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.0500g/mL
Deuterium (D) 98.5% min.
Flash Point 20°C
Packaging Glass Bottle
Refractive Index 1.506
Water 0.1% max. (Coulometric)
Quantity 5 mL
Melting Point -41.0°C
Boiling Point 114.4°C
Beilstein 20, III, 2305
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5, 99 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine-d5
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'