missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d{5}, 100%(Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051974
Click to view available options
Quantity:
10 Ea.
2 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.05 g/mL
Flash Point 20°C (68°F)
Odor Unpleasant
Packaging Prescored Ampule
Refractive Index 1.506
Quantity 2 Ea.
Melting Point -42°C
Boiling Point 115°C
UN Number UN1282
Beilstein 114377
Formula Weight 84.14
Percent Purity 100%
Chemical Name or Material Pyridine-d5, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'