missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ACS reagent, Honeywell

Catalog Number p-7073756
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1
This item is not returnable. View return policy
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1
UN Number UN1282
Beilstein 103233
Formula Weight 79.1g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density 0.978g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Vapor Pressure 10 mmHg (13.2°C), 20 mmHg (25°C)
Chemical Name or Material Pyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'