missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
100 mL
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
pH | 8.8 (at 20°C) |
Melting Point | -42°C |
Color | Colorless |
UN Number | UN1282 |
Beilstein | 103233 |
Solubility Information | Completely miscible in water |
Formula Weight | 79.1g/mol |
Percent Purity | ≥99% |
Density | 0.978g/mL at 25°C |
Boiling Point | 115°C |
Show More |
Safety and Handling
Product Identifier
- Pyridine
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : pyridine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'