missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 214.07 |
Percent Purity | 95% |
Chemical Name or Material | Pyridine-3-sulfonyl chloride hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'