missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-3-carboxamidine hydrochloride 0.5 hydrate, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Assay Percent Range | 95% |
Quantity | 10 g |
Formula Weight | 166.61 |
Percent Purity | 95% |
Chemical Name or Material | Pyridine-3-carboxamidine hydrochloride 0.5 hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'