missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 10658034
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1692-25-7
C5H6BNO2
122.92
MFCD00674177
ABMYEXAYWZJVOV-UHFFFAOYSA-N
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
2734378
OB(O)C1=CC=CN=C1
This item is not returnable. View return policy
1692-25-7
C5H6BNO2
122.92
MFCD00674177
ABMYEXAYWZJVOV-UHFFFAOYSA-N
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
2734378
OB(O)C1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name (pyridin-3-yl)boronic acid
Formula Weight 122.92
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Pyridine-3-boronic acid
compliance-icons
Product Identifier
  • Pyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'