missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboximidamide hydrochloride, 97%, Maybridge™

£13.06 - £155.00

Chemical Identifiers

CAS 51285-26-8
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00052271
InChI Key GMHCEDDZKAYPLB-UHFFFAOYSA-N
Synonym picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
PubChem CID 12403615
IUPAC Name pyridine-2-carboximidamide;hydrochloride
SMILES C1=CC=NC(=C1)C(=N)N.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10069251
SDS View Documents
Maybridge
RF01042DA
1g Amber Glass Bottle
£13.06
1g
View alternative products
Save Up to 
»
Estimated Shipment
14-04-2021
Please to purchase this item. Need a web account? Register with us today!
10242452
SDS View Documents
Maybridge
RF01042EA
10g Amber Glass Bottle
£71.33
10g
View alternative products
Save Up to 
»
Estimated Shipment
14-04-2021
Please to purchase this item. Need a web account? Register with us today!
10614395
SDS View Documents
Maybridge
RF01042EB
25g Amber Glass Bottle
£155.00
25g
View alternative products
Save Up to 
»
Estimated Shipment
14-04-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

51285-26-8
157.601
GMHCEDDZKAYPLB-UHFFFAOYSA-N
12403615
C1=CC=NC(=C1)C(=N)N.Cl
C6H8ClN3
MFCD00052271
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
pyridine-2-carboximidamide;hydrochloride

Specifications

Pyridine-2-carboximidamide hydrochloride
1g
97.0
C6H8ClN3
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
C1=CC=NC(=C1)C(=N)N.Cl
157.601
157.6
Amber Glass Bottle
51285-26-8
97%
MFCD00052271
GMHCEDDZKAYPLB-UHFFFAOYSA-N
pyridine-2-carboximidamide;hydrochloride
12403615
97%
SDS