Learn More
Pyridine-2-carboximidamide hydrochloride, 97%, Maybridge™
Brand: Maybridge RF01042DA
Code : 31
Additional Details : CAS Number : 51285-26-8 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
51285-26-8 | |
157.601 | |
GMHCEDDZKAYPLB-UHFFFAOYSA-N | |
12403615 | |
C1=CC=NC(=C1)C(=N)N.Cl |
C6H8ClN3 | |
MFCD00052271 | |
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride | |
pyridine-2-carboximidamide;hydrochloride |
Specifications
Pyridine-2-carboximidamide hydrochloride | |
1g | |
97.0 | |
C6H8ClN3 | |
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride | |
C1=CC=NC(=C1)C(=N)N.Cl | |
157.601 | |
157.6 |
Amber Glass Bottle | |
51285-26-8 | |
97% | |
MFCD00052271 | |
GMHCEDDZKAYPLB-UHFFFAOYSA-N | |
pyridine-2-carboximidamide;hydrochloride | |
12403615 | |
97% |