missing translation for 'onlineSavingsMsg'
Learn More

PYREX™ Shorty Disposable Glass Serological Pipets

Ideal for use in confined areas such as hoods

Brand:  Corning 7077B-1

Code : A0

Additional Details : Weight : 5.66990kg

 View more versions of this product

Product Code. 10716323

  • £247.82 / Pack of 500


Estimated Shipment
23-02-2018
Add to basket

Description

Description

  • Sterile, plugged pipets have tooled tops
  • Negative graduations make it applicable for both serological and measuring procedures
  • I.S.O. color coding is printed directly on Steri-View™ (one side paper, one side plastic) wrapper

Specifications

Specifications

Yes
To Deliver/Blow Out
Yellow
Yes
215mm
Individually Wrapped
Tooled
1mL
Sterile
Yes
0.03 oz., 1mL
Yes
0.01mL
0.5mL
Round
Serological Pipet
Pyrex™
Videos