missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Screwcap Spare Inserts

For use with Black SVL™ PBT screwcaps

Brand:  Pyrex 4513/60M

Code : 01

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 11393484

  • £68.90 / Pack of 5
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

  • PTFE faced sealing disc for chemical resistance
  • Autoclavable up to 121°C
Specifications

Specifications

60 mm
Spare insert
5 Pack
Autoclavable
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers