missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key

For use when liquids have to be added to systems under vacuum.

Brand:  Pyrex 443540

Code : NEW

Additional Details : Weight : 0.47000kg

 View more versions of this product

Product Code. 13431869

  • £95.33 / Each


In stock, ready to ship: 6
Add to basket

Description

Description

  • Complies with ISO 4800, Type 4
  • With interchangeable PTFE key for grease free performance
  • Double graduated scale showing volume of liquid contained/delivered
  • Stem inside cone for easy fitting to Quickfit™ assemblies
  • Graduations in white enamel
  • Supplied with hollow glass stopper
Specifications

Specifications

Yes
Pressure Equalizing Tube
Borosilicate Glass
10mL
100 mL
Videos
SDS
Documents