Learn More
Pyrex™ Narrow-Mouth Reagent Bottle
Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.
Brand: Pyrex 1526/04D
Code : A0
Additional Details : Weight : 1.14000kg
Description
Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.
- Unground neck for use with rubber stoppers (not supplied)
- Borosilicate glass provides strong chemical resistance
- Complies with ISO 4796
Specifications
ISO 4796 | |
54 mm | |
95 mm | |
100 mL | |
10pack | |
Yes |
No | |
Rubber Stoppers | |
Narrow | |
Borosilicate Glass | |
Reagent Bottle | |
Unground |