missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Büchner Flask with Screw Thread Connector

Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.

Brand:  Pyrex 1170/02SC

Code : 01

Additional Details : Weight : 0.12596kg

 View more versions of this product

Product Code. 15117203

  • £17.95 / Each


In Stock 7
Add to basket

Description

Description

  • Complies with ISO 6556
  • Büchner flasks with heavy wall construction for vacuum filtration applications
  • sidearm features a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing
  • With tooled neck to accept a drilled rubber stopper
Specifications

Specifications

ISO 6556, DIN 12476
64mm
Borosilicate Glass
PTFE screw thread connector
24mm (Neck I.D.)
100mL
105mm
Videos