Learn More
Pyrex™ Round Bottom Threaded Culture Tubes
Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.
Brand: Corning 99449-20X
Code : 02
Additional Details : Weight : 12.40000kg
Description
- Can be reliably used in almost any assay procedure from cell to chemical analysis
- Made from chemically inert Pyrex® borosilicate glass
- Uniform base and wall thickness reduce the risk of breakage due to thermal shock and prevents warping or bending
- Phenolic style screw caps available (separately) with PTFE liner for autoclave sterilisation, or rubber liner for hot air sterilisation
- Supplied in convenient shelf pack quantities
Specifications
30mL | |
20mm | |
20mm | |
Borosilicate Glass | |
Yes | |
Ideal for use with samples sensitive to leaching from plastic tubes | |
Bulk | |
Untreated | |
18-415 |
Screw neck | |
Yes | |
150mm | |
No | |
No | |
None | |
Round | |
No | |
Round |