missing translation for 'onlineSavingsMsg'
Learn More

PYREX™ Disposable Round-Bottom Rimless Glass Tubes

Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.

Brand:  Corning 99445-18

Code : 02

Additional Details : Weight : 9.88042kg

 View more versions of this product

Product Code. 10735763

  • £105.00 / Pack of 500


In Stock 23
Add to basket

Description

Description

 

  • Packaged in easy-to-open and easy-to-use bulk packs
  • Not treated

 

Specifications

Specifications

28.5mL
18mm
Borosilicate Glass
Yes
For tissue culture work and general bacteriological use
Bulk
Untreated
Round
18mm
150mm
No
No
None
Round
No
Videos