missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYREX™ Disposable Rimless Flat-Bottom Glass Tubes
Ideal for plant tissue culture and general culture work
Brand: Corning 9850-25
Code : Z1
Additional Details : Weight : 0.80000kg
Description
- Walls and bottoms are uniform thickness; flat bottom facilitates use with agar slants
- Tubes are rimless for greater ease in plugging and rack storage
- Two-position polypropylene closures available separately; position closure OPEN for gas exchange or CLOSED for humidified environment
- Internal drip ring minimizes contamination
Specifications
39mL | |
100mm | |
No | |
No | |
None | |
Untreated |
25mm | |
Borosilicate Glass | |
No | |
For tissue culture work and general bacteriological use | |
Flat | |
No |