missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Round Bottom Threaded Culture Tubes

Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.

Brand:  Corning 99449-20

Code : A0

Additional Details : Weight : 11.24908kg

 View more versions of this product

Product Code. 10631854

  • £244.39 / Pack of 500


Estimated Shipment
28-02-2018
Add to basket

Description

Description

  • Can be reliably used in almost any assay procedure from cell to chemical analysis
  • Made from chemically inert Pyrex® borosilicate glass
  • Uniform base and wall thickness reduce the risk of breakage due to thermal shock and prevents warping or bending
  • Phenolic style screw caps available (separately) with PTFE liner for autoclave sterilisation, or rubber liner for hot air sterilisation
  • Supplied in convenient shelf pack quantities
Specifications

Specifications

24mL
20mm
Without
125mm
No
No
None
Round
18-415
Screw neck
Yes
20mm
Borosilicate Glass
Yes
Ideal for use with samples sensitive to leaching from plastic tubes
Bulk
Untreated
Round
Videos