Learn More
Pyrex™ Borosilicate Glass Watch Glasses
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
Brand: Pyrex 1060/10D
Code : 01
Additional Details : Weight : 0.02068kg
Description
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
- For evaporation work and covering heated beakers
- Manufactured from heat resisting Pyrex™ borosilicate glass
- Fire-polished edges to reduce chipping and breakage
Specifications
Flat | |
Round | |
Evaporation Work and Covering Heated Beakers | |
80 mm |
Borosilicate Glass | |
Watch Glass | |
Polished |