missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Vacuum Filter Funnel with Glass Sintered Disc
Useful when conventional filter papers react with chemicals.
Brand: Quickfit VB60/06M
102.60 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.18900kg
Description
- Büchner vacuum filtration funnel with glass sintered disc
- Manufactured from borosilicate glass for highest resistance to chemical attack
- Useful where conventional filter papers would be destroyed by acids and alkalis
- With a Quickfit™ ground glass cone to allow direct filtration into Quickfit™ Büchner flasks
Specifications
ISO 3585, DIN 12217, ASTM E-438 | |
125 mL | |
Borosilicate Glass | |
Quickfit™ ground Glass cone |