missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Round Bottom Threaded Culture Tubes

Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.

Brand:  Corning 99449-16XX

Code : 02

Additional Details : Weight : 18.76000kg

 View more versions of this product

Product Code. 13023083

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Can be reliably used in almost any assay procedure from cell to chemical analysis
  • Made from chemically inert Pyrex® borosilicate glass
  • Uniform base and wall thickness reduce the risk of breakage due to thermal shock and prevents warping or bending
  • Phenolic style screw caps available (separately) with PTFE liner for autoclave sterilisation, or rubber liner for hot air sterilisation
  • Supplied in convenient shelf pack quantities
Specifications

Specifications

19mL
16mm
Without
150mm
No
No
None
Round
No
Round
Screw neck
Yes
16mm
Borosilicate Glass
Yes
Ideal for use with samples sensitive to leaching from plastic tubes
Bulk
Untreated
15-415
Videos
Documentation View accessory products