missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pyrene-1-boronic acid, 95%

Brand:  Alfa Aesar™ H64211.06

Code : NEW

Additional Details : CAS Number : 164461-18-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401319

  • £115.00 / 5g

Estimated Shipment: 04-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

164461-18-1
246.07
MWEKPLLMFXIZOC-UHFFFAOYSA-N
5084102
OB(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
C16H11BO2
MFCD04974062
1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid, boronic acid, 1-pyrenyl, 1-pyrene boronic acid, pyreneboronic acid, pubchem16490, pyren-1-yl boronic acid
pyren-1-ylboronic acid

Specifications

Pyrene-1-boronic acid
247°C to 251°C (literature)
164461-18-1
MFCD04974062
1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid, boronic acid, 1-pyrenyl, 1-pyrene boronic acid, pyreneboronic acid, pubchem16490, pyren-1-yl boronic acid
MWEKPLLMFXIZOC-UHFFFAOYSA-N
pyren-1-ylboronic acid
5084102
Powder
Yellow
5g
C16H11BO2
7134073
Slightly soluble in water.
OB(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
246.07
246.07
95%
Videos
SDS Documents
Certificates