missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 434340050

Code : 29

Additional Details : CAS Number : 378211-85-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10536630

  • £549.40 / 5g


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
≥96.0%
C7H5N3O2
MFCD00297221
378211-85-9
97%
163.14

Specifications

Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
Authentic
Powder or Crystalline Powder
97%
163.14
White to orange
378211-85-9
Glass bottle
≥96.0%
C7H5N3O2
MFCD00297221
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pyrazolo¬Ę1,5-a!pyrimidine-2-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS