missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2704901
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16205-46-2
C8H6N2O2
162.148
MFCD00229214
HRSDPDBQVZHCRC-UHFFFAOYSA-N
pyrazolo 1,5-a pyridine-3-carboxylic acid, zlchem 84, 3-pyrazolo 1,5-a pyridinecarboxylic acid, pyrazolo 1,5-a pyridine-3-carboxylicacid, pyrazolo pyridine-3-carboxylic acid, h-pyrazolo 1,5-a pyridine-3-carboxylic acid, 8-hydropyrazolo 1,5-a pyridine-3-carboxylic acid
2795462
pyrazolo[1,5-a]pyridine-3-carboxylic acid
C1=CC2=C(C=NN2C=C1)C(=O)O
This item is not returnable. View return policy
16205-46-2
C8H6N2O2
162.148
MFCD00229214
HRSDPDBQVZHCRC-UHFFFAOYSA-N
pyrazolo 1,5-a pyridine-3-carboxylic acid, zlchem 84, 3-pyrazolo 1,5-a pyridinecarboxylic acid, pyrazolo 1,5-a pyridine-3-carboxylicacid, pyrazolo pyridine-3-carboxylic acid, h-pyrazolo 1,5-a pyridine-3-carboxylic acid, 8-hydropyrazolo 1,5-a pyridine-3-carboxylic acid
2795462
pyrazolo[1,5-a]pyridine-3-carboxylic acid
C1=CC2=C(C=NN2C=C1)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Pyrazolo[1, 5-a]pyridine-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'