Pyrazinecarbonitrile, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 436920500

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 19847-12-2 Weight: 0.10000kg Transport: UN number : 3276 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 50g

Product Code. 10735441

Quantity Price
1 £ 67.6 / 50g
Estimated Shipment date
from Supplier 18-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Pyrazinecarbonitrile
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H3N3
Formula Weight 105.1

Specification

Formula Weight 105.1
Physical Form Liquid
Assay ≥98.5%
Quantity 50g
Color Clear colorless to yellow
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 19847-12-2
Refractive Index 1.5330 to 1.5350 (20°C, 589nm)
Assay Percent Range 99%
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C5H3N3
Density 1.174g/mL
Chemical Name or Material Pyrazinecarbonitrile
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.