missing translation for 'onlineSavingsMsg'
Learn More

Pyramex™ Lanyard

Catalog Number 11823480
Click to view available options
Product Type:
Retaining Cord w/ Plastic Clips
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in retaining spectacles

Fits a variety of temples.
Color Black
Product Type Retaining Cord w/ Plastic Clips
For Use With (Application) Retaining spectacles
Includes Plastic clips
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'