missing translation for 'onlineSavingsMsg'
Learn More

PX 12, Tocris Bioscience™

Thioredoxin-1 inhibitor

Brand:  Tocris (Bio-Techne) 295450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462124

  • £440.72 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Thioredoxin-1 (Trx-1) inhibitor; irreversibly inactivates Trx-1 as a substrate for reduction by thioredoxin reductase (TR); competitively inhibits the reduction of Trx-1 by TR (Ki = 30.8μM). Attenuates expression of HIF-1α, VEGF and iNOS (IC50 values are 7.2, 10.4 and 18.1μM respectively). Exhibits antitumor activity; directly inhibits tubulin in vitro and decreases tumor microvessel density in vivo.
Specifications

Chemical Identifiers

141400-58-0
188.307
2-sec-butyldisulfanyl-1h-imidazole, 2-1-methylpropyl dithio-1h-imidazole, 2-butan-2-yldisulfanyl-1h-imidazole, 2-1-methylpropyl dithio-1h-imidazole, 1h-imidazole,2-1-methylpropyl dithio, 1h-imidazole, 2-1-methylpropyl dithio, 1h-imidazole, 2-1-methylpropyl dithio, iv 2 compound, iv-2 compound, 1-methylpropyl-2-imidazolyl disulfide
2-(butan-2-yldisulfanyl)-1H-imidazole
C7H12N2S2
BPBPYQWMFCTCNG-UHFFFAOYSA-N
219104
CCC(C)SSC1=NC=CN1

Specifications

PX 12
141400-58-0
2-sec-butyldisulfanyl-1h-imidazole, 2-1-methylpropyl dithio-1h-imidazole, 2-butan-2-yldisulfanyl-1h-imidazole, 2-1-methylpropyl dithio-1h-imidazole, 1h-imidazole,2-1-methylpropyl dithio, 1h-imidazole, 2-1-methylpropyl dithio, 1h-imidazole, 2-1-methylpropyl dithio, iv 2 compound, iv-2 compound, 1-methylpropyl-2-imidazolyl disulfide
CCC(C)SSC1=NC=CN1
188.307
188.31
50mg
C7H12N2S2
BPBPYQWMFCTCNG-UHFFFAOYSA-N
2-(butan-2-yldisulfanyl)-1H-imidazole
219104
>99%
Videos