Allsigns International™ PVC Safety Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PHT9

Safety tape PHT9. photoluminescent PVC selfadhesive 80mm x 10m fire exit right running

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12319737

Quantity Price
1 £ 318.75 / Each
Estimated Shipment
03-03-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Adhesive Self-adhesive
Product Type Safety Tape
For Use With (Application) Highlighting hazards/restricted areas
Color White, Photoluminescent
Material PVC
Dimensions (L x W) 80mm x 10m
Documentation

Technical documents