Allsigns International™ PVC Safety Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PHT7

Safety tape PHT7. photoluminescent PVC selfadhesive white 80mm x 10m Allsigns, 1/Cs, MA: TBD

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12399727

Quantity Price
1 £ 318.75 / Each
Estimated Shipment
03-03-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
For Use With (Application) Highlighting hazards/restricted areas
Adhesive Self-adhesive
Color White, Photoluminescent
Material PVC
Product Type Safety Tape
Dimensions (L x W) 80mm x 10m
Documentation

Technical documents