Allsigns International™ PVC Safety Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PHT11

Safety tape PHT11. photoluminescent PVC selfadhesive 80mm x 10m fire exit left running

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12339737

Quantity Price
1 £ 318.75 / Each
Estimated Shipment
28-02-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
For Use With (Application) Highlighting hazards/restricted areas
Adhesive Self-adhesive
Color Photoluminescent
Material PVC
Product Type Safety Tape
Dimensions (L x W) 10m x 80mm
Documentation

Technical documents