Allsigns International™ PVC Safety Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PHT6

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12389727

Quantity Price
1 £ 188.71 / Each
Estimated Shipment
06-06-2017
Add to basket


Description and Specification

Specification

Material PVC
Dimensions (L x W) 10m x 40mm
Color White, Photoluminescent
Quantity 1Each
For Use With (Application) Highlighting hazards/restricted areas
Adhesive Self-adhesive
Product Type Safety Tape
Documentation

Technical documents