Allsigns International™ PVC Safety Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PHT10

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12329737

Quantity Price
1 £ 188.71 / Each
Estimated Shipment
09-06-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Highlighting hazards/restricted areas
Color Photoluminescent
Quantity 1Each
Product Type Safety Tape
Adhesive Self-adhesive
Material PVC
Dimensions (L x W) 40mm x 10m
Documentation

Technical documents