ALLSIGNS International™ PVC Safety Sign


Overview
Brand: ALLSIGNS International™

Manufacturer Part Number: SAP-785-K

Safety sign SA21/PVC/R. fire exit with runningman/directional arrow right pointing arrow 750mm

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12399807

Quantity Price
1 £ 34.63 / Each
Estimated Shipment
14-03-2017
Add to basket


Description and Specification

Specification

Size (Metric) 750 x 150mm
Quantity 1Each
Type Safety Sign
Material PVC
Message Fire Exit with Running Man/Directional Arrow Right Pointing Arrow
Documentation

Technical documents