ALLSIGNS International™ PVC Safety Sign


Overview
Brand: ALLSIGNS International™

Manufacturer Part Number: MA9

Safety sign T2/PVC mandatory sign this area mustbe kept clear at all times 200mm x 300mm 1mm

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12389937

Quantity Price
1 £ 21.21 / Each
Estimated Shipment
01-03-2017
Add to basket


Description and Specification

Specification

Size (Metric) 200 x 300mm
Quantity 1Each
Type Safety Sign
Material PVC
Message This Area Must be kept Clear at All Times
Documentation

Technical documents