missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Safety Sign

Brand:  Allsigns International™ EN8PHR

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12319867

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Description

Gemglow self adhesive backed 1mm rigid plastic 82mm x 202mm photoluminescent fire hose reel sign Allsigns
Description & Specifications

Specifications

82mm
Fire hose reel sign
Safety Sign
202mm
Documentation

Technical Documents