Allsigns International™ PVC Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: EN9PH

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12329867

Quantity Price
1 £ 9.95 / Each
Estimated Shipment
06-04-2017
Add to basket


Description and Specification

Specification

Message Fire extinguisher sign powder M28
Dimensions (L x W) 82mm x 202mm
Type Safety Sign

Photoluminescent fire extinguisher sign Gemglow self adhesive backed 1mm rigid plastic 82mm x 202mm powder M28 Allsigns

Documentation

Technical documents