missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Safety Sign

Brand:  Allsigns International™ EN10PHR

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12339867

  • POA / Each


Estimated Shipment
02-08-2017
Add to basket

Specifications

Description

Photoluminescent fire extinguisher sign Gemglow self adhesive backed 1mm rigid plastic 82mm x 202mm B.C.F Allsigns
Description & Specifications

Specifications

Fire extinguisher sign B.C.F
Safety Sign
82mm x 202mm
Documentation

Technical Documents