Allsigns International™ PVC Reflective Floor Marking Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: REF2

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12359727

Quantity Price
1 £ 299.39 / Each
Estimated Shipment
09-05-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Product Type Reflective Floor Marking Tape
Dimensions (L x W) 50mm x 25m
Color Black and Yellow, Photoluminescent
Material PVC
Adhesive Self-adhesive
For Use With (Application) Highlighting hazards/restricted areas
Documentation

Technical documents