missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International Safety tape REF2. 50mm x 25m photoluminescent zebra

Brand:  Allsigns International REF2

Safety tape REF2. 50mm x 25m photoluminescent zebra

Code : 04

Additional Details : Weight : 0.01000kg

Product Code. 12369727

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »